Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

651885

Sigma-Aldrich

Dicyclohexyl(2-methylphenyl)phosphine

95%

Synonym(s):

(o-Tolyl)dicyclohexylphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H29P
CAS Number:
Molecular Weight:
288.41
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

assay

95%

form

solid

reaction suitability

reaction type: Cross Couplings

mp

90-93 °C

functional group

phosphine

SMILES string

Cc1ccccc1P(C2CCCCC2)C3CCCCC3

InChI

1S/C19H29P/c1-16-10-8-9-15-19(16)20(17-11-4-2-5-12-17)18-13-6-3-7-14-18/h8-10,15,17-18H,2-7,11-14H2,1H3

InChI key

MKHYBAIPMJGEQO-UHFFFAOYSA-N

General description

Dicyclohexyl(2-methylphenyl)phosphine is a phosphorus ligand with some unique features like high melting point, ease of handle and less susceptibility for oxidation, making it an ideal catalyst in coupling reactions.[1]

Application

Cocatalyst for:
  • Nickel-catalyzed coupling reactions of alkenes, aldehydes, and silyl triflates

Catalyst for:
  • Coupling of nonactivated and deactivated aryl chlorides with arylboronic acid
  • Preparation of (alkyl)(aryl)phosphines and triarylphosphines, as mediators of rhodium-catalyzed propene and hexene hydroformylation reactions

  • Ligand for PC bond cleavage during Heck vinylation of aryl halides with Bu acrylate in presence of palladium catalyst compounds
Dicyclohexyl(2-methylphenyl)phosphine [PCy2(o-Tol)] may be used along with [IrCl(cod)]2 to form a complex that can catalyze the addition of aliphatic acid chlorides to terminal alkynes to form (Z)-β-chloro-α,β-unsaturated ketones.[2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Armarego WLF and Chai CLL
Purification of Laboratory Chemicals, 736-736 (2013)
Iridium-catalyzed addition of aroyl chlorides and aliphatic acid chlorides to terminal alkynes.
Iwai T, et al.
Journal of the American Chemical Society, 134(2), 1268-1274 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service