Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

651508

Sigma-Aldrich

(1R,2R)-Pseudoephedrine, polymer-bound

50-100 mesh, extent of labeling: 1.0-2.0 mmol/g loading, 1 % cross-linked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

form

solid

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

1.0-2.0 mmol/g loading

particle size

50-100 mesh

functional group

amine

storage temp.

2-8°C

Application

Used for asymmetric alkylations on solid phase.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Panee C Hutchison et al.
The Journal of organic chemistry, 69(3), 790-801 (2004-01-31)
A pseudoephedrine resin has been successfully employed in asymmetric alkylations on solid phase. Immobilized pseudoephedrine amides are conveniently prepared by the one-step attachment of pseudoephedrine to Merrifield resin through the hydroxyl group and subsequent acylation on nitrogen. Deprotonation and alkylation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service