Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

650838

Sigma-Aldrich

(S)-(+)-1-Benzyloxy-2-propanol

97%

Synonym(s):

(S)-1-O-Benzylpropylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

assay

97%

optical activity

[α]20/D +14.5°, c = 1 in chloroform

refractive index

n20/D 1.510 (lit.)

density

1.044 g/mL at 25 °C (lit.)

SMILES string

C[C@H](O)COCc1ccccc1

InChI

1S/C10H14O2/c1-9(11)7-12-8-10-5-3-2-4-6-10/h2-6,9,11H,7-8H2,1H3/t9-/m0/s1

InChI key

KJBPYIUAQLPHJG-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(+)-1-Benzyloxy-2-propanol can be used as an effective antiwear agent in aviation turbine fuels, diesel fuels, and light mineral oil.[1]

Application

(S)-(+)-1-Benzyloxy-2-propanol can be used as a reactant to prepare:
  • (S)-N-(2-phosphonomethoxypropyl) derivatives of purine and pyrimidine bases (PMP derivatives).[1]
  • (S)-1-(benzyloxy)propan-2-yl 4-methylbenzenesulfonate by reacting with tosyl chloride in pyridine.[2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

227.8 °F

flash_point_c

108.8 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-catalyzed cross-coupling of nonactivated secondary alkyl halides and tosylates with secondary alkyl Grignard reagents
Yang C-T, et al.
Journal of the American Chemical Society, 134(27), 11124-11127 (2012)
Enantiomeric impurities in chiral catalysts, auxiliaries, and synthons used in enantioselective syntheses. Part 4
Qiu H, et al.
Tetrahedron Asymmetry, 24(18), 1134-1141 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service