Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

650161

Supelco

Methanol solution

suitable for HPLC, contains Methanol:Acetonitrile 1:1 (v/v)

Synonym(s):

Acetonitrile:Methanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH4O
CAS Number:
Molecular Weight:
32.04
Beilstein/REAXYS Number:
3662861
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
technique(s):
HPLC: suitable
bp:
65.4 °C (lit.)
vapor pressure:
410 mmHg

Product Name

Methanol solution, (Methanol:Acetonitrile 1:1 (v/v))

vapor density

1.11 (vs air)

Quality Level

vapor pressure

410 mmHg

form

liquid

expl. lim.

5.5-44 % (lit.)

concentration

(Methanol:Acetonitrile 1:1 (v/v))

technique(s)

HPLC: suitable

bp

65.4 °C (lit.)

mp

-98 °C (lit.)

density

0.787 g/mL at 20 °C

format

mixture

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

4×4 L packaged in glass bottles; 18 L packaged in VerSA-Flow containers

Legal Information

VerSA-Flow is a trademark of Sigma-Aldrich Co. LLC

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

41.0 °F - closed cup

flash_point_c

5 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Åsa Björkbacka et al.
Dalton transactions (Cambridge, England : 2003), 44(36), 16045-16051 (2015-08-20)
One of the main challenges for the nuclear power industry today is the disposal of spent nuclear fuel. One of the most developed methods for its long term storage is the Swedish KBS-3 concept where the spent fuel is sealed
Takashi Fujii et al.
Nature protocols, 10(9), 1445-1456 (2015-08-28)
Live single-cell mass spectrometry (live MS) provides a mass spectrum that shows thousands of metabolite peaks from a single live plant cell within minutes. By using an optical microscope, a cell is chosen for analysis and a metal-coated nanospray microcapillary
Danielle Hagstrom et al.
Toxicological sciences : an official journal of the Society of Toxicology, 147(1), 270-285 (2015-06-28)
Traditional toxicology testing has relied on low-throughput, expensive mammalian studies; however, timely testing of the large number of environmental toxicants requires new in vitro and in vivo platforms for inexpensive medium- to high-throughput screening. Herein, we describe the suitability of the asexual

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service