Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

649686

Sigma-Aldrich

Poly(bis(1,4-dioxapentyl)phosphazene)

Synonym(s):

Poly(bis(2-methoxyethoxy)phosphazene)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H14NO4P)n
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

λmax

225 nm

General description

Poly [(organo) phosphazene] polymers has an inorganic skeletal structure consisting of alternating nitrogen and phosphorous atoms connected by alternating single and double bonds.[1]

Application

Poly (bis(2-methoxyethoxy) phosphazene may be used to prepare graft copolymers with poly(ε-caprolactone) to be used in potential drug delivery and tissue engineering applications.[2]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Applicative aspects of poly(organophosphazenes) (2004)
Synthesis and Characterization of Poly (bis (2-methoxyethoxy) phosphazene)-graft-Poly (epsilon-caprolactone) Copolymers.
Deng L, et al.
Acta Polymerica Sinica / Gao Fen Zi Xue Bao, 1, 88-88 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service