64955
(−)-B-Methoxydiisopinocampheylborane hydrate
technical
Synonym(s):
(−)-Diisopinocampheylmethoxyborane
About This Item
Recommended Products
grade
technical
form
solid
optical activity
[α]20/D −72±3°, c = 1% in diethyl ether
impurities
≤2% water
SMILES string
COB([C@@H]1CC2CC([C@H]1C)C2(C)C)[C@H]3CC4CC([C@H]3C)C4(C)C
InChI
1S/C21H37BO/c1-12-16-8-14(20(16,3)4)10-18(12)22(23-7)19-11-15-9-17(13(19)2)21(15,5)6/h12-19H,8-11H2,1-7H3/t12-,13-,14+,15+,16-,17-,18-,19?/m1/s1
InChI key
IAQXEQYLQNNXJC-CLIXPIQASA-N
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service