Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

649155

Sigma-Aldrich

6-Methoxypyridine-2-boronic acid N-phenyldiethanolamine ester

Synonym(s):

6-Methoxy-2-pyridineboronic acid-N-phenyldiethanolamine ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19BN2O3
CAS Number:
Molecular Weight:
298.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

mp

210 °C (dec.) (lit.)

SMILES string

COC1=CC=CC(B(OCC2)OCCN2C3=CC=CC=C3)=N1

InChI

1S/C16H19BN2O3/c1-20-16-9-5-8-15(18-16)17-21-12-10-19(11-13-22-17)14-6-3-2-4-7-14/h2-9H,10-13H2,1H3

InChI key

DBLBCHOTUMUCCT-UHFFFAOYSA-N

General description

May contain varying amounts of Isopropanol and N-phenyldiethanolamine as stabilizers

application

Reactant for:
  • Preparation of alkylated sulfonamides via alkylation with alcohols catalyzed by Ag/Mo hybrid
  • Preparation of 7-disubstituted oxyindoles as EP3 receptor antagonists
  • Preparation of N-substituted sulfonamides via ferrous chloride-catalyzed N-alkylation with benzylic alcohols
  • Preparation of (alkyl)hydroxybenzimidazoles, via a O- to N-acyl transfer reaction, as intermediates for EP3 receptor antagonists
  • Preparation of N-arylsulfonyl β-[(aryloxy)indolyl]acrylamides as potent and selective EP3 receptor antagonists

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service