Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

649007

Sigma-Aldrich

3,4-Dimethyl-N-(4-fluorobenzylidene)aniline

97%

Synonym(s):

N-(4-Fluorobenzylidene)-3,4-xylidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH=NC6H3(CH3)2
CAS Number:
Molecular Weight:
227.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

49-53 °C (lit.)

functional group

fluoro
imine

SMILES string

Cc1ccc(cc1C)\N=C\c2ccc(F)cc2

InChI

1S/C15H14FN/c1-11-3-8-15(9-12(11)2)17-10-13-4-6-14(16)7-5-13/h3-10H,1-2H3/b17-10+

Inchi Key

HBEHYGAVDFVSNN-LICLKQGHSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

>229.8 °F

flash_point_c

> 109.9 °C

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service