Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

64875

Sigma-Aldrich

Bis(dimethylamino)methoxymethane

technical

Synonym(s):

Methoxy-bis(dimethylamino)methane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH[N(CH3)2]2
CAS Number:
Beilstein/REAXYS Number:
1900972
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

refractive index

n20/D 1.418

bp

120-125 °C (lit.)

density

0.863 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(N(C)C)N(C)C

InChI

1S/C6H16N2O/c1-7(2)6(9-5)8(3)4/h6H,1-5H3

Inchi Key

NCIMAYPZWJQYGN-UHFFFAOYSA-N

Other Notes

Review[1]

pictograms

Flame

signalword

Danger

hcodes

pcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

50.0 °F - closed cup

flash_point_c

10 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R.F. Abdulla et al.
Tetrahedron, 35, 1675-1675 (1979)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service