Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

648671

Sigma-Aldrich

Ytterbium-172Yb(III) oxide

95 atom % (172Yb)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
172Yb2O3
Molecular Weight:
391.87

isotopic purity

95 atom % (172Yb)

form

solid

SMILES string

[172Yb][172Yb](=O)(=O)=O

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service