Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

646628

Sigma-Aldrich

Poly(5-(3,7-dimethyloctyloxy)-2-methoxy-cyanoterephthalylidene)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C40H54N2O4)n
CAS Number:
MDL number:
UNSPSC Code:
12352103

form

solid

semiconductor properties

N-type (mobility~10−5 cm2/V·s)

General description

Light emitting conjugated polymer.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The development of high-performance conjugated organic molecules and polymers has received widespread attention in industrial and academic research.

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service