646512
Methyl 4-(3-thienyl)benzoate
97%
Synonym(s):
Methyl 4-(thiophen-3-yl)benzoate
About This Item
assay
97%
form
solid
mp
160-164 °C (lit.)
SMILES string
COC(=O)c1ccc(cc1)-c2ccsc2
InChI
1S/C12H10O2S/c1-14-12(13)10-4-2-9(3-5-10)11-6-7-15-8-11/h2-8H,1H3
Inchi Key
NTBKCEONBLOPPY-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service