Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

646261

Sigma-Aldrich

1,3-Diethyl-1,1,3,3-tetramethyldisilazane

>98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H23NSi2
CAS Number:
Molecular Weight:
189.45
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

>98.0%

form

liquid

bp

37-39 °C/0.2 mmHg (lit.)

density

0.817 g/mL at 25 °C (lit.)

SMILES string

CC[Si](C)(C)N[Si](C)(C)CC

InChI

1S/C8H23NSi2/c1-7-10(3,4)9-11(5,6)8-2/h9H,7-8H2,1-6H3

InChI key

MHRNQQUEUYMEEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

104.0 °F - closed cup

flash_point_c

40 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

atomic layer deposition (ALD), microelectronics, Mo:Al2O3 films, nanocomposite coating, photovoltaics, semiconductor devices, W:Al2O3 films, composite films, layer-by-layer

Synthesis of Melting Gels Using Mono-Substituted and Di-Substituted Alkoxysiloxanes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service