Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

646172

Sigma-Aldrich

1-(4-Methylbenzyl)piperazine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N2
CAS Number:
Molecular Weight:
190.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

37-41 °C (lit.)

SMILES string

Cc1ccc(CN2CCNCC2)cc1

InChI

1S/C12H18N2/c1-11-2-4-12(5-3-11)10-14-8-6-13-7-9-14/h2-5,13H,6-10H2,1H3

InChI key

RNAXUUAJNMDESG-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

204.8 °F - closed cup

flash_point_c

96 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service