Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

645834

Sigma-Aldrich

3-Bromo-2-(2′-fluorobenzyloxy)phenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11BBrFO3
CAS Number:
Molecular Weight:
324.94
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

mp

116-120 °C (lit.)

SMILES string

OB(O)c1cccc(Br)c1OCc2ccccc2F

InChI

1S/C13H11BBrFO3/c15-11-6-3-5-10(14(17)18)13(11)19-8-9-4-1-2-7-12(9)16/h1-7,17-18H,8H2

InChI key

XTBIXNSHJQLMFR-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service