Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

645214

Sigma-Aldrich

Hexane

95%, purification grade

Synonym(s):

n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein/REAXYS Number:
1730733
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
grade:
purification grade
assay:
95%
bp:
69 °C (lit.)
vapor pressure:
256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)
Pricing and availability is not currently available. Contact Technical Service

Product Name

Hexane, purification grade, 95%

grade

purification grade

vapor density

~3 (vs air)

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

assay

95%

form

liquid

autoignition temp.

453 °F

expl. lim.

7.7 %

evapn. residue

≤0.0003%

refractive index

n20/D 1.375 (lit.)

bp

69 °C (lit.)

mp

−95 °C (lit.)

density

0.659 g/mL at 25 °C (lit.)

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-7.6 °F - closed cup

flash_point_c

-22 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Russell J Chedgy et al.
Phytochemistry, 113, 149-159 (2015-01-07)
Salicinoids are phenolic glycosides (PGs) characteristic of the Salicaceae and are known defenses against insect herbivory. Common examples are salicin, salicortin, tremuloidin, and tremulacin, which accumulate to high concentrations in the leaves and bark of willows and poplars. Although their

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service