Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

644447

Sigma-Aldrich

Hexane, mixture of isomers

purification grade

Synonym(s):

Hexanes, Isohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18
Beilstein/REAXYS Number:
1730733
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
purification grade
bp:
68-70 °C (lit.)
Pricing and availability is not currently available. Contact Technical Service

Product Name

Hexane, mixture of isomers, purification grade

grade

purification grade

vapor density

3 (vs air)

autoignition temp.

437 °F

expl. lim.

7.5 %

evapn. residue

≤0.0005%

refractive index

n20/D 1.379 (lit.)

bp

68-70 °C (lit.)

mp

−95 °C (lit.)

solubility

ethanol: soluble(lit.)

density

0.672 g/mL at 25 °C (lit.)

InChI

1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3

InChI key

AFABGHUZZDYHJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-9.4 °F

flash_point_c

-23 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 492-492 (1994)
Separation of hexane isomers in a metal-organic framework with triangular channels.
Herm ZR, et al.
Science, 340(6135), 960-964 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service