Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

643742

Sigma-Aldrich

trans-Bromo(N-succinimidyl)bis(triphenylphosphine)palladium(II)

97%

Synonym(s):

Bromobis(triphenylphosphine)(N-succinimide)palladium(II), [Pd(NCOC2H4CO)(PPh3)2Br]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H34BrNO2P2Pd
CAS Number:
Molecular Weight:
808.98
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

223-228 °C (dec.) (lit.)

SMILES string

Br[Pd]N1C(=O)CCC1=O.c2ccc(cc2)P(c3ccccc3)c4ccccc4.c5ccc(cc5)P(c6ccccc6)c7ccccc7

InChI

1S/2C18H15P.C4H5NO2.BrH.Pd/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;6-3-1-2-4(7)5-3;;/h2*1-15H;1-2H2,(H,5,6,7);1H;/q;;;;+2/p-2

InChI key

KYQYWUJRFOCJEW-UHFFFAOYSA-L

Application

trans-Bromo(N-succinimidyl)bis(triphenylphosphine)palladium(II) may be used in the following processes:
  • As a catalyst for the preparation of 6-cyano-2-(3-fluoro-benzyl)-indole-1-carboxylic acid tert-butyl ester via Suzuki cross-coupling reaction between 1-boc-6-cyanoindole-2-boronic acid and 3-fluorobenzyl bromide. This method does not involve the use of strong bases or toxic reagents and also the use of this catalyst minimizes protodeboronation.[1]
  • As a precatalyst for the Suzuki-Miyaura cross-coupling reaction between alkenyl tosylates and alkenyl MIDA boronates in the absence of phosphine ligands. For less activated alkenyl tosylate, the addition of tricyclohexylphosphine tetrafluoroborate improves the reactivity.[2]
Catalyst for C-C bond formation, e.g. Stille coupling reaction and Suzuki coupling.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Burns et al.
Organic letters, 9(26), 5397-5400 (2007-12-01)
trans-PdBr(N-Succ)(PPh3)2 (1) is a universally effective precatalyst for Suzuki-Miyaura cross-couplings of benzylic halides with aryl- or heteroarylboronic acids. Substituted aryl halides and halogenated cyclic enones can be cross-coupled with aryl- or vinylboronic acids in excellent yields. Catalyst recycling is also
Suzuki-Miyaura cross-coupling of alkenyl tosylates with alkenyl MIDA boronates.
Luthy M and Taylor RJK.
Tetrahedron Letters, 53(27), 3444-3447 (2012)
Palladium-catalyzed benzylation of N-Boc indole boronic acids.
Kearney AM, et al.
Tetrahedron Letters, 51(17), 2281-2283 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service