About This Item
Recommended Products
form
solid
reaction suitability
core: rhodium
reagent type: catalyst
extent of labeling
5 wt. % dry loading
weight
Unit excludes wt. of H2O
matrix
carbon, wet support
SMILES string
[Rh]
InChI
1S/Rh
InChI key
MHOVAHRLVXNVSD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service