Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

642614

Sigma-Aldrich

1-Isobutylpyrazole-4-boronic acid pinacol ester

95%

Synonym(s):

1-Isobutyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-Isobutyl-pyrazole-4-boronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H23BN2O2
CAS Number:
Molecular Weight:
250.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

95%

refractive index

n20/D 1.4740 (lit.)

bp

248-249 °C (lit.)

density

1.003 g/mL at 25 °C (lit.)

SMILES string

CC(C)Cn1cc(cn1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C13H23BN2O2/c1-10(2)8-16-9-11(7-15-16)14-17-12(3,4)13(5,6)18-14/h7,9-10H,8H2,1-6H3

InChI key

YMEBZRNYQBODKB-UHFFFAOYSA-N

Application

1-Isobutylpyrazole-4-boronic acid pinacol ester can be used:
  • In the synthesis of 7-azaindole derivatives as potent PDK1 inhibitors.[1]
  • As a reactant in the synthesis of imidazo[1,2-a]pyrazine derivatives as potent phosphodiesterase 10A inhibitors.[2]
  • To prepare 2-substituted-3-aroyl-benzo[b]furan derivatives.[3]

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

116.6 °F

flash_point_c

47 °C

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient synthesis and substitution of 3-aroyl-2-bromobenzo [b] furans
Gill GS, et al.
The Journal of Organic Chemistry, 73(3), 1131-1134 (2008)
Discovery of novel 7-azaindoles as PDK1 inhibitors
Wucherer-Plietker M, et al.
Bioorganic & medicinal chemistry letters, 26(13), 3073-3080 (2016)
Discovery of a potent, selective, and orally active phosphodiesterase 10A inhibitor for the potential treatment of schizophrenia
Bartolome?-Nebreda JM, et al.
Journal of Medicinal Chemistry, 57(10), 4196-4212 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service