Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

642185

Sigma-Aldrich

trans-2-Chloro-beta-nitrostyrene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH=CHNO2
CAS Number:
Molecular Weight:
183.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

45-49 °C (lit.)

functional group

amine
chloro
nitro

SMILES string

[H]\C(=C(\[H])[N+]([O-])=O)c1ccccc1Cl

InChI

1S/C8H6ClNO2/c9-8-4-2-1-3-7(8)5-6-10(11)12/h1-6H/b6-5+

InChI key

QHKJTRDWAZGBLR-AATRIKPKSA-N

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service