Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

641898

Sigma-Aldrich

1,8-Diaminonaphthalene-d10

96 atom % D, 97% D (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D10N2
Molecular Weight:
168.26
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

96 atom % D
97% D (CP)

form

solid

bp

205 °C/12 mmHg (lit.)

mp

60-65 °C (lit.)

mass shift

M+10

SMILES string

[2H]N([2H])c1c([2H])c([2H])c([2H])c2c([2H])c([2H])c([2H])c(N([2H])[2H])c12

InChI

1S/C10H10N2/c11-8-5-1-3-7-4-2-6-9(12)10(7)8/h1-6H,11-12H2/i1D,2D,3D,4D,5D,6D/hD4

InChI key

YFOOEYJGMMJJLS-XBXNOAEOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Health hazardExclamation mark

signalword

Warning

pcodes

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service