Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

641448

Sigma-Aldrich

N-Acetamidoacrylic acid, polymer-bound

100-200 mesh, extent of labeling: 1.0-1.5 mmol/g loading, 1 % cross-linked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

form

solid

crosslinking

1 % cross-linked with divinylbenzene

extent of labeling

1.0-1.5 mmol/g loading

particle size

100-200 mesh

storage temp.

2-8°C

Application

Polymer-bound reagent for α-substituted amino acid and indole synthesis.,

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbaste, M.
Tetrahedron Letters, 39, 6287-6287 (1998)
Kondo, Y.
Journal of the Chemical Society. Perkin Transactions 1, 2137-2137 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service