Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

640697

Sigma-Aldrich

(1E,3Z)-1-Methoxy-2-methyl-3-(trimethylsilyloxy)-1,3-pentadiene

Synonym(s):

(1E,3Z)-1-Methoxy-2-methyl-3-[(trimethylsilyl)oxy]-penta-1,3-diene, [(1-(2-Methoxy-1-methylvinyl)propenyloxy]trimethylsilan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O2Si
CAS Number:
Molecular Weight:
200.35
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

refractive index

n20/D 1.4650 (lit.)

bp

65 °C/6 mmHg (lit.)

density

0.899 g/mL at 25 °C (lit.)

functional group

ether

storage temp.

2-8°C

SMILES string

CO\C=C(C)\C(O[Si](C)(C)C)=C\C

InChI

1S/C10H20O2Si/c1-7-10(9(2)8-11-3)12-13(4,5)6/h7-8H,1-6H3/b9-8+,10-7-

InChI key

SSGAQVOLXCOJGN-AZRUORMVSA-N

Application

Danishefsky diene used as a reactant for:
Preparation of migrastatin analog as cell migration inhibitor for treatment of tumor metastasis
Preparation of dihydropyridinones by chlorosilane-promoted aza-Diels-Alder addition
Synthesis on non-proteinogenic amino esters via stereoselective Lewis acid-catalyzed aza-Diels-Alder cycloaddition with conjugated (cyclo)dienes

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

178.0 °F - closed cup

flash_point_c

81.1 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service