Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

640433

Sigma-Aldrich

2-Chloro-2-ethoxyacetate, polymer-bound

50-100 mesh, extent of labeling: 0.50-1.50 mmol/g loading, 1 % cross-linked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.22

form

solid

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

0.50-1.50 mmol/g loading

particle size

50-100 mesh

storage temp.

2-8°C

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymer-supported alpha-imino acetates. Versatile reagents for the synthesis of alpha-amino acid libraries.
S Kobayashi et al.
Journal of combinatorial chemistry, 2(5), 438-440 (2000-10-13)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service