640093
6-(Trifluoromethyl)pyridine-3-carbonitrile
97%
Synonym(s):
6-(Trifluoromethyl)nicotinonitrile
About This Item
Recommended Products
assay
97%
form
solid
mp
37-41 °C (lit.)
SMILES string
FC(F)(F)c1ccc(cn1)C#N
InChI
1S/C7H3F3N2/c8-7(9,10)6-2-1-5(3-11)4-12-6/h1-2,4H
InChI key
ZBLULFJVRGVQAW-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 2 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
180.0 °F - closed cup
flash_point_c
82.2 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service