All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H3Cl2FO2S
CAS Number:
Molecular Weight:
229.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
assay
95%
refractive index
n20/D 1.5550 (lit.)
bp
243-244 °C (lit.)
mp
27-31 °C
density
1.569 g/mL at 25 °C (lit.)
SMILES string
Fc1ccc(c(Cl)c1)S(Cl)(=O)=O
InChI
1S/C6H3Cl2FO2S/c7-5-3-4(9)1-2-6(5)12(8,10)11/h1-3H
InChI key
FCFPJKHVCHKCMP-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
>230.0 °F - closed cup
flash_point_c
> 110 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service