Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

637009

Sigma-Aldrich

Poly(2,5-didodecylphenylene-1,4-ethynylene)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

solubility

chlorinated solvents: soluble
toluene: soluble

Looking for similar products? Visit Product Comparison Guide

General description

Conjugated polymer.

Application

For optical and electronic uses

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Materials Design Concepts for Efficient Blue OLEDs: A Joint Theoretical and Experimental Study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service