About This Item
Recommended Products
assay
97%
form
solid
mp
45-50 °C (lit.)
functional group
chloro
SMILES string
CC(C)(C)OC(=O)n1ccc2cc(Cl)ccc12
InChI
1S/C13H14ClNO2/c1-13(2,3)17-12(16)15-7-6-9-8-10(14)4-5-11(9)15/h4-8H,1-3H3
InChI key
GBBULIKZPZSTLA-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Chronic 4 - Skin Sens. 1
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service