Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

636746

Sigma-Aldrich

3-Chloro-4-pyridinecarboxaldehyde

97%

Synonym(s):

3-Chloroisonicotinaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4ClNO
CAS Number:
Molecular Weight:
141.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

solid

mp

58-62 °C (lit.)

SMILES string

[H]C(=O)c1ccncc1Cl

InChI

1S/C6H4ClNO/c7-6-3-8-2-1-5(6)4-9/h1-4H

InChI key

ZVGDKOQPJCOCLI-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheuk-Fai Chow et al.
Chemistry, an Asian journal, 3(8-9), 1324-1335 (2008-07-30)
Nine neutral dynamic metallosupramolecular polymers (metallodynamers) based on acyl hydrazone derived metal coordination centers (Co(2+), Ni(2+), Zn(2+), and Cd(2+)) were generated through self-assembly polymerization. These are linear coordination polymers with specific optical and mechanical properties. Monomer selection was found to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service