Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

636045

Sigma-Aldrich

2-(Methanesulfonylamino)phenylboronic acid pinacol ester

97%

Synonym(s):

N-[2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20BNO4S
CAS Number:
Molecular Weight:
297.18
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

130-134 °C (lit.)

SMILES string

CC1(C)OB(OC1(C)C)c2ccccc2NS(C)(=O)=O

InChI

1S/C13H20BNO4S/c1-12(2)13(3,4)19-14(18-12)10-8-6-7-9-11(10)15-20(5,16)17/h6-9,15H,1-5H3

InChI key

ADAOINPTCZWNBZ-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service