Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

635553

Sigma-Aldrich

PAMAM-sodium carboxylate dendrimer, 1,6-diaminohexane core, generation 3.5 solution

10 wt. % in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)6NH2]:(G=3.5);dendri PAMAM(NHCH2CH2COONa)64
Molecular Weight:
12983.80
MDL number:
UNSPSC Code:
12162002

description

sodium carboxylate surface groups

form

solution

mol wt

generation 3.5

feature

core type 1,6-diaminohexane core (6-carbon core)

concentration

10 wt. % in methanol

no. Surface Groups

64

storage temp.

−20°C

Features and Benefits

Contains 64 surface sodium carboxylate groups.
Dendrimers with sodium salt surfaces will exhibit higher solubility in more polar organic or aqueous systems. These particles will be particularly useful where specific surface charges are beneficial or where sodium salts may promote transport to other properties

Packaging

unit weight includes solvent

Legal Information

Mfg. by Dendritic NanoTechnologies Inc. Licensed for research use only.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service