Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

634859

Sigma-Aldrich

3-Methoxy-2-naphthol

97%

Synonym(s):

2-Hydroxy-3-methoxynaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC10H6OH
CAS Number:
Molecular Weight:
174.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

107-111 °C (lit.)

SMILES string

COc1cc2ccccc2cc1O

InChI

1S/C11H10O2/c1-13-11-7-9-5-3-2-4-8(9)6-10(11)12/h2-7,12H,1H3

InChI key

SJTNTIRIRIPZDV-UHFFFAOYSA-N

Application

3-Methoxy-2-naphthol can be used in the synthesis of a naphthoate ester, which can act like dopamine D4 antagonist.[1]

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design and synthesis of 2-naphthoate esters as selective dopamine D4 antagonists.
I Boyfield et al.
Journal of medicinal chemistry, 39(10), 1946-1948 (1996-05-10)
Nick D Pokorzynski et al.
eLife, 8 (2019-04-03)
During infection, pathogens are starved of essential nutrients such as iron and tryptophan by host immune effectors. Without conserved global stress response regulators, how the obligate intracellular bacterium Chlamydia trachomatis arrives at a physiologically similar 'persistent' state in response to
Nenad Manevski et al.
Drug metabolism and disposition: the biological fate of chemicals, 43(1), 126-139 (2014-10-24)
Although skin is the largest organ of the human body, cutaneous drug metabolism is often overlooked, and existing experimental models are insufficiently validated. This proof-of-concept study investigated phase II biotransformation of 11 test substrates in fresh full-thickness human skin explants

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service