Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

634778

Sigma-Aldrich

4-Hydroxyiodobenzene diacetate, polymer-bound

100-200 mesh, extent of labeling: 0.5-1.5 mmol/g I loading, 1 % cross-linked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

form

solid

crosslinking

1 % cross-linked with divinylbenzene

extent of labeling

0.5-1.5 mmol/g I loading

particle size

100-200 mesh

Application

Polymer-bound oxidant.

pictograms

Exclamation mark

signalword

Warning

hcodes

pcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service