Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

634360

Sigma-Aldrich

Nickel cobalt oxide

nanopowder, <150 nm particle size (BET), 99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiO CoO
CAS Number:
Molecular Weight:
192.56
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

assay

99% trace metals basis

form

nanopowder

particle size

<150 nm (BET)

density

6.6 g/mL at 25 °C (lit.)

SMILES string

O=[Co].O=[Ni]

InChI

1S/Co.Ni.2O

InChI key

YTBWYQYUOZHUKJ-UHFFFAOYSA-N

Legal Information

Product of Engi-Mat Co.

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 Inhalation

target_organs

Lungs

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Magnetic materials find diverse applications from data storage to renewable energy.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service