Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

633836

Sigma-Aldrich

Strontium ferrite

nanopowder, <100 nm particle size (BET), 99.8% trace metals basis

Synonym(s):

Iron strontium oxide, Strontium hexaferrite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrFe12O19
CAS Number:
Molecular Weight:
1061.75
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

assay

99.8% trace metals basis

form

crystalline (hexagonal phase)
nanopowder

particle size

<100 nm (BET)

mp

>450 °C (lit.)

density

5.18 g/mL at 25 °C (lit.)

SMILES string

O=[Sr].O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O

InChI

1S/12Fe.19O.Sr

InChI key

MUDURESJCZWWBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service