Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

633828

Sigma-Aldrich

Barium strontium titanium oxide

nanopowder, <100 nm particle size (APS), >99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(BaTiO3)(SrTiO3)
Molecular Weight:
416.68
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

>99% trace metals basis

form

nanopowder

particle size

<100 nm (APS)

density

4.91 g/mL at 25 °C (lit.)

SMILES string

O1[Sr]O[Ti]12O[Ti]3(O[Ba]O3)O2

InChI

1S/Ba.6O.Sr.2Ti

InChI key

WIUXIYWRELFPAN-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huifang Zhao et al.
ACS applied materials & interfaces, 10(43), 37732-37742 (2018-10-09)
It is challenging to realize a dual-ion mode of matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOF MS) for detecting small molecules. Herein, graphene coated by porous amorphous carbon with P-O surface group and codoped by phosphorus and nitrogen (O-P,N-C/G) was

Articles

Synthesis, Properties, and Applications of Perovskite-Phase Metal Oxide Nanostructures

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Among various ceramics, one-dimensional (1-D) piezoelectric ceramics have attracted significant scientific attention for use in energy harvesting.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service