Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

633461

Sigma-Aldrich

4-Formyl-3-hydroxybenzoic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6O4
CAS Number:
Molecular Weight:
166.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

mp

235-240 °C (lit.)

functional group

aldehyde
carboxylic acid

SMILES string

OC(=O)c1ccc(C=O)c(O)c1

InChI

1S/C8H6O4/c9-4-6-2-1-5(8(11)12)3-7(6)10/h1-4,10H,(H,11,12)

InChI key

FDDHFCWYCKQKGY-UHFFFAOYSA-N

Application

4-Formyl-3-hydroxybenzoic acid can be used as a reactant to prepare:
  • A pH-sensitive fluorescent probe 4,4′-(hydrazine-1,2-diylidene bis(methanylylidene)) bis(3-hydroxybenzoic acid (HDBB) by one-step condensation reaction with hydrazine.[1]
  • 2-Oxo-2H-1-benzopyran-3,7-dicarboxylic acid by condensation reaction with diethyl malonate.[2]
  • Schiff base ligands for the preparation of stable and functional Schiff base metal complexes.[3][4]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple route to dinuclear complexes containing unusual μ-N sulfonamido bridges
Sanmartin-Matalobos, et al.
Journal of Coordination Chemistry, 69(8), 1358-1370 (2016)
Synthesis of some new benzocoumarin heterocyclic fluorescent dyes
Shahinian Edward GH and Sebe I
Revista de Chimie, 62, 1098-1101 (2011)
Palladium (ii) complexes of tetradentate donor-acceptor Schiff base ligands: synthesis and spectral, structural, thermal and NLO properties
Celedon S, et al.
New. J. Chem., 44(22), 9190-9201 (2020)
An aggregation-induced emission-based pH-sensitive fluorescent probe for intracellular acidity sensing
Lin Na, et al.
Royal Society of Chemistry Advances, 6(30), 25416-25419 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service