Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

633445

Sigma-Aldrich

6-Quinolinyl trifluoromethanesulfonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6F3NO3S
CAS Number:
Molecular Weight:
277.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

34-38 °C (lit.)

SMILES string

FC(F)(F)S(=O)(=O)Oc1ccc2ncccc2c1

InChI

1S/C10H6F3NO3S/c11-10(12,13)18(15,16)17-8-3-4-9-7(6-8)2-1-5-14-9/h1-6H

InChI key

TVYXNKUMLWUQPV-UHFFFAOYSA-N

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service