Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

632783

Sigma-Aldrich

2-Acetamido-5-aminopyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N3O
CAS Number:
Molecular Weight:
151.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

97%

mp

154-158 °C (lit.)

SMILES string

CC(=O)Nc1ccc(N)cn1

InChI

1S/C7H9N3O/c1-5(11)10-7-3-2-6(8)4-9-7/h2-4H,8H2,1H3,(H,9,10,11)

InChI key

SXGURHSLXPDJBC-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Linear free energy approach to the substituent effects in ultraviolet absorption spectra of pyridines. I. Mono-and disubstituted pyridines"
Tomasik.P and Zakowicz.A
Chemical Papers, 31(02), 254-264 (1977)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service