Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

632503

Sigma-Aldrich

Poly(tetrahydrofurfuryl methacrylate)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

General description

Poly(tetrahydrofurfuryl methacrylate) is biocompatible in nature. The structure of the polymer remains rigid even after absorbing water up to 75%.[1] Large diffusion of water results in lowering of glass transition temperature.[2]

Application

Poly(tetrahydrofurfuryl methacrylate) may find potential uses in medical implants.[2]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physicochemical Behavior and Supramolecular Organization of Polymers (2009)
P D Riggs et al.
Biomaterials, 20(5), 435-441 (1999-04-16)
Poly(tetrahydrofurfuryl methacrylate) possesses some unique characteristics with respect to its biocompatibility and behaviour in water. The water uptake is high (>70%) and very slow (over 3 yr), but the material remains rigid throughout the process. The mechanism behind the uptake

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service