Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

632449

Supelco

Water solution

contains 0.10 % (v/v) trifluoroacetic acid, 10.0 % (v/v) methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

form

liquid

contains

0.10 % (v/v) trifluoroacetic acid
10.0 % (v/v) methanol

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

4×4 L packaged in glass bottles; 20 L packaged in NOWPAK HDPE Bottle; 18 L packaged in VerSA-Flow containers

Preparation Note

Product filtered through a 0.2 μm filter

Legal Information

VerSA-Flow is a trademark of Sigma-Aldrich Co. LLC

signalword

Warning

pcodes

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - STOT SE 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

140.0 °F - closed cup

flash_point_c

60 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service