Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

631159

Sigma-Aldrich

3-Bromo-N-methylbenzylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10BrN
CAS Number:
Molecular Weight:
200.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.5650 (lit.)

bp

231-232 °C (lit.)

density

1.461 g/mL at 25 °C (lit.)

functional group

amine
bromo

SMILES string

CNCc1cccc(Br)c1

InChI

1S/C8H10BrN/c1-10-6-7-3-2-4-8(9)5-7/h2-5,10H,6H2,1H3

InChI key

QCEANFBGRBLXHN-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service