Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

630640

Sigma-Aldrich

Benzyl phenyl carbonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCO2C6H5
CAS Number:
Molecular Weight:
228.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.5490 (lit.)

bp

120-130 °C/0.5 mmHg (lit.)

density

1.156 g/mL at 25 °C (lit.)

SMILES string

O=C(OCc1ccccc1)Oc2ccccc2

InChI

1S/C14H12O3/c15-14(17-13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10H,11H2

InChI key

SNGLYCMNDNOLOF-UHFFFAOYSA-N

General description

Benzyl phenyl carbonate can be synthesized by reacting phenyl chloroformate with benzyl alcohol in the presence of pyridine.[1]

Application

Benzyl phenyl carbonate can be used to synthesize carbamate protected polyamines.[1]

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

?Selective synthesis of carbamate protected polyamines using alkyl phenyl carbonates"
Pittelkow M, et al.
Synthesis, 2002(15), 2195-2202 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service