Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

630179

Sigma-Aldrich

Poly(2,2,3,3,4,4,4-heptafluorobutyl acrylate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C7H5F7O2)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

contains

0.1 mol % modified with 1-Dodecyl mercaptan (based on total monomer)

refractive index

n20/D 1.377 (lit.)
n20/D 1.3770 (lit.)

transition temp

Tg -30 °C(lit.)

solubility

DMSO: soluble
NMP: soluble
THF: soluble
methylene chloride: soluble

density

1.498 g/mL at 25 °C (lit.)

General description

Transparent, low surface tension polymer.

Application

Low refractive index polymer for cladding layer in optical waveguides.

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

85.0 °F - closed cup

flash_point_c

29.44 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service