Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

630160

Sigma-Aldrich

Poly(2,2,3,4,4,4-hexafluorobutyl acrylate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C7H6F6O2)n
MDL number:
UNSPSC Code:
12352103

refractive index

n20/D 1.394 (lit.)

transition temp

Tg -22 °C(lit.)

density

1.519 g/mL at 25 °C (lit.)

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

182.0 °F - closed cup

flash_point_c

83.33 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service