Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

616893

Sigma-Aldrich

Phenyl o-xylyethane-d18

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

98 atom % D

application(s)

electronics

mass shift

M+18

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])C([2H])([2H])c2c([2H])c([2H])c([2H])c(c2C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C16H18/c1-13-7-6-10-16(14(13)2)12-11-15-8-4-3-5-9-15/h3-10H,11-12H2,1-2H3/i1D3,2D3,3D,4D,5D,6D,7D,8D,9D,10D,11D2,12D2

InChI key

SZFXZCSXSJOEAM-UOZRLOLDSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service