Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

615765

Sigma-Aldrich

Methyl benzoate-d8

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5CO2CD3
Molecular Weight:
144.20
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

bp

198-199 °C (lit.)

mp

-12 °C (lit.)

density

1.152 g/mL at 25 °C

mass shift

M+8

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C(=O)OC([2H])([2H])[2H]

InChI

1S/C8H8O2/c1-10-8(9)7-5-3-2-4-6-7/h2-6H,1H3/i1D3,2D,3D,4D,5D,6D

InChI key

QPJVMBTYPHYUOC-JGUCLWPXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

181.4 °F - closed cup

flash_point_c

83.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronald V Emmons et al.
Journal of separation science, 43(9-10), 1915-1924 (2020-02-27)
The continued rise in the extraction of unconventional oil and gas across the globe poses many questions about how to manage these relatively new waste-streams. Produced water, the primary waste by-product, contains a diverse number of anthropogenic additives together with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service