Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

613495

Sigma-Aldrich

1-Amino(octane-d17)

98 atom % D

Synonym(s):

Octylamine-d17

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)7NH2
CAS Number:
Molecular Weight:
146.35
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

assay

99% (CP)

bp

179 °C (lit.)

mp

0-1 °C (lit.)

density

0.883 g/mL at 25 °C

mass shift

M+17

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])N

InChI

1S/C8H19N/c1-2-3-4-5-6-7-8-9/h2-9H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2

InChI key

IOQPZZOEVPZRBK-OISRNESJSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

140.0 °F - closed cup

flash_point_c

60 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service