Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

612308

Sigma-Aldrich

Dimethyl sulfoxide-d6, Extra

Extra, ≥99.996 atom % D

Synonym(s):

Dimethyl sulfoxide-d6, (Methyl sulfoxide)-d6, DMSO-d6, Hexadeuterodimethyl sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2SO
CAS Number:
Molecular Weight:
84.17
Beilstein/REAXYS Number:
1237248
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

vapor pressure

0.42 mmHg ( 20 °C)

Quality Level

isotopic purity

≥99.996 atom % D

assay

≥99.0%

autoignition temp.

573 °F

expl. lim.

42 %

impurities

≤0.025% water
water

refractive index

n20/D 1.476 (lit.)

bp

189 °C (lit.)

mp

20.2 °C (lit.)

density

1.190 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3/i1D3,2D3

InChI key

IAZDPXIOMUYVGZ-WFGJKAKNSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

190.4 °F

flash_point_c

88 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service